missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 4N solution in 1,4-Dioxane/water, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 366521000

35.65 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.



 View more versions of this product

Product Code. 10464272

  • £47.24 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless to yellow liquid
Colorless to Yellow
7647-01-0, 7732-18-5, 123-91-1
1.1000g/mL
1.1
100 mL
HCl
MFCD00011324 MFCD00792839
04,450; 05,533; 06,283
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Hydrochloric acid
4N Solution in 1, 4-dioxane/water
3.95 to 4.40N
Glass bottle
12°C
ClH
Liquid
2924
15, 4818
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.45
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid, 4N solution in 1,4-Dioxane/water

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only