missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 5% v/v aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1 L
4 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Yellow |
Concentration | 5% v/v aq. soln. |
Quantity | 500 mL |
Physical Form | Liquid |
UN Number | UN1789 |
IUPAC Name | dichloropalladium |
Formula Weight | 36.46 |
Chemical Name or Material | Hydrochloric acid |
Safety and Handling
Product Identifier
- Hydrochloric acid, 5% v/v aqueous solution
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrochloric acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'