missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid, 6N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™ Product Code.: 15599660
compliance-icons

Hydrochloric Acid, 6N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15599660
1 L, Narrow Mouth Polyethylene Bottle
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Polyethylene Bottle
This item is not returnable. View return policy

Product Code. 15599660

Brand: Avantor J.T.Baker 561902

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.10kg
Quantity 1 L
Formula Weight 36.46
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Hydrochloric Acid, 6N Volumetric Solution
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid, 6N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.