missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 99.99% Trace Metals Basis, Honeywell™
37 wt. % in H2O, 99.999% trace metals basis
£94.33 - £325.10
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15674100
|
Honeywell Research Chemicals
339253-100ML |
100 mL | Poly Bottle |
£94.33
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15684100
|
Honeywell Research Chemicals
339253-500ML |
500 mL | Poly Bottle |
£325.10
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
>0.1 (at 20°C) | |
7647-01-0 | |
42°C | |
Stinging | |
ClH | |
MFCD00011324 MFCD00792839 | |
1098214 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46g/mol | |
Hydrochloric acid |
Colorless | |
-35°C | |
1.2g/mL at 25°C | |
3.23psi (21.1°C), 7.93psi (37.7°C) | |
Liquid | |
UN1789 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
99.999% (trace metals basis) |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
missing translation for 'dotInformation' : Class 8/ PGII
missing translation for 'rtecsNumber' : 231-595-7
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrochloric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid