missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, 99.999% (metals basis), 36.5% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1000 mL
2.5 L
Description
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Appearance | Colorless liquid |
Color | Colorless |
Boiling Point | 81.5°C to 110°C |
Density | 1.19 g/mL |
Odor | Pungent |
Assay Percent Range | 99.999% (metals basis) |
Quantity | 250 mL |
Physical Form | Liquid |
Solubility Information | Completely miscible with water. |
Formula Weight | 36.46 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'