missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, for analysis, fuming, 37% solution in water

Product Code. 10755663 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10755663

Brand: Thermo Scientific Acros 124630100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0, 7732-18-5
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Melting Point -35.0°C
Boiling Point 57.0°C
Density 1.1800g/mL
Heavy Metals (as Pb) 2ppm max.
Packaging Plastic drum
Reducing Substances (SO2) Passes Test
Specific Gravity 1.18
Quantity 10 L
Viscosity 1.9 mPa.s (15°C)
Linear Formula HCl
Physical Form Solution
Fieser 04,450; 05,533; 06,283
Solubility Information Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
IUPAC Name chlorane
Formula Weight 36.46
Percent Purity 35.0 to 39.0%
Grade Analytical
Chemical Name or Material Hydrochloric acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.