missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Calibration and Matrix Blank For ICP, SPEX CertiPrep™
Click to view available options
Quantity:
500 mL
Packaging:
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Hydrochlorid Acid Matrix
- 500 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
CAS | 7647-01-0 |
Molecular Formula | ClH |
MDL Number | MFCD00011324 MFCD00792839 |
Synonym | Muriatic Acid |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
SMILES | Cl |
IUPAC Name | hydrogen chloride |
Molecular Weight (g/mol) | 36.46 |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
Show More |
Product intended for laboratory use only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'