missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric acid concentrate, For 1L standard solution, 0.2 M HCl (0.2N), Honeywell Fluka™ Product Code.: 15635280

Hydrochloric acid concentrate, For 1L standard solution, 0.2 M HCl (0.2N), Honeywell Fluka™

Product Code. 15635280 Shop All Honeywell Fluka Products
1 Pc.
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy

Product Code. 15635280

Brand: Honeywell-Fluka 382871EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 1L standard solution, 0.2 M HCl (0.2N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Packaging Ampoule
Vapor Pressure 190 mmHg (25°C)
Quantity 1 Pc.
UN Number UN1789
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid concentrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric acid concentrate, For 1L standard solution, 0.2 M HCl (0.2N), Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.