missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid NF/FCC/ACS, Macron Fine Chemicals™
Click to view available options
Quantity:
500 mL
2.5 L
4 x 2.5 L
Packaging:
Glass
Glass Bottle
SAFEMOR
Specifications
Specifications
Packaging | Glass |
Quantity | 4 x 2.5 L |
IUPAC Name | hydrogen chloride |
Formula Weight | 36.46 |
Grade | ACS/FCC/NF |
Chemical Name or Material | Hydrochloric Acid, (HCl 36.5%-38.0%) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction