missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid NF/FCC/ACS, Macron Fine Chemicals™

Product Code. 15587944
Click to view available options
Quantity:
500 mL
2.5 L
4 x 2.5 L
Packaging:
Glass
Glass Bottle
SAFEMOR
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

Packaging SAFEMOR
Quantity 500 mL
IUPAC Name hydrogen chloride
Formula Weight 36.46
Grade ACS/FCC/NF
Chemical Name or Material Hydrochloric Acid, (HCl 36.5%-38.0%)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'