missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, puriss. p.a., Honeywell Fluka™
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
25 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | Colorless |
Density | 1.16g/mL at 20°C, 1.2g/mL at 25°C |
Vapor Pressure | 3.23 psi (21.1°C), 7.93 psi (37.7°C) |
Quantity | 250 mL |
UN Number | UN1789 |
Beilstein | 1098214 |
IUPAC Name | hydrogen chloride |
Formula Weight | 36.46g/mol |
Percent Purity | ≥32% (T) |
Grade | puriss. p.a. |
Show More |
Safety and Handling
Product Identifier
- Hydrochloric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'