missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid R1, , Ph Eur Grade, Reagecon™
BIOPHARMA

Product Code. 11464689
Click to view available options
Quantity:
1 L
Packaging:
1L
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Packaging 1L
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • hydrochloric acid r1
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H312-Harmful in contact with skin.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P360-Rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'