missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Ready-to-use standardized solution 0.5M, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
Specifications
CAS | 7647-01-0, 7732-18-5 |
Concentration | 0.5M (0.5N) |
Packaging | Bottle |
Quantity | 1000 mL |
Molecular Formula | ClH |
MDL Number | MFCD00011324 MFCD00792839 |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
SMILES | Cl |
IUPAC Name | hydrogen chloride |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'