missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
124.95 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 Ampule
6 Ampules
Description
ClH, CAS Number-7647-01-0, spirits of salt, hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride, chlorwasserstoff, hydrogen chloride hcl, muriatic acid, anhydrous hydrochloric acid, acide chlorhydrique, 6AMP, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Boiling Point | 100°C |
Packaging | Ampoule |
Quantity | 6 Ampules |
Physical Form | Liquid |
Formula Weight | 36.46g/mol |
Chemical Name or Material | Hydrochloric Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'