missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Meets Analytical Spec. Of Ph.Eur., Bp

ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol

£29.90 - £50.90

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10325710
View accessory products View Documents
Fisher Chemical
J/4350/15
1 L
£29.90
1 L
Please to purchase this item. Need a web account? Register with us today!
10419560
View accessory products View Documents Promotion Details
Fisher Chemical
J/4350/17
2.5 L
£50.90
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ClH, CAS Number-7647-01-0, chlorohydric acid, chlorwasserstoff, anhydrous hydrochloric acid, acide chlorhydrique, chloorwaterstof, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 2.5L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

2
7647-01-0
HDPE plastic bottle
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46g/mol
Colorless
100°C
14mmHg @20°C
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid, 0.1M, (0.1N)

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

SDS
Documents

Documents

Product Certifications