missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ Product Code.: 12616727

Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 12616727
25 L, Plastic container
Click to view available options
Quantity:
5 L
10 L
25 L
Packaging:
Bag in box
Plastic container
This item is not returnable. View return policy

Product Code. 12616727

Brand: Fisher Chemical J/4350/25

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ClH, CAS Number-7647-01-0, hydrogen chloride, hydrochloric acid, acide chlorhydrique, chlorwasserstoff, anhydrous hydrochloric acid, chlorohydric acid, chloorwaterstof, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 100 deg.C, CHEBI:17883, Colorless, 36.46g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstofShow MoreShow Less
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

pH 2
Color Colorless
Boiling Point 100°C
Packaging Plastic container
Vapor Pressure 14mmHg @20°C
Quantity 25 L
Physical Form Liquid
IUPAC Name chlorane
Formula Weight 36.46g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2526652 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog NumberJ/4350/21,J/4350C/90,J/4350/25
compliance-icons
Product Identifier
  • Hydrochloric acid 0.1M
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid Solution 0.1M (0.1N), NIST Standard Solution Ready To Use, for Volumetric Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.