missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Description
Hydrochloric Acid Solution 0.5M (0.5N), NIST Standard Solution Ready To Use, for Volumetric Analysis
Specifications
Specifications
Color | Colorless |
Boiling Point | 100°C |
Packaging | Plastic container |
Quantity | 25 L |
Physical Form | Liquid |
Formula Weight | 36.46g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'