missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 0.5M, Honeywell Fluka™

Catalog Number 15693530
Click to view available options
Quantity:
500 mL
2 L
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
chlorane
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
chlorane
Cl

Volumetric, 0.5 M HCl (0.5N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY
Concentration 0.5M
Density 1.01g/mL at 25°C
Vapor Pressure 190mmHg (25°C)
Quantity 500 mL
UN Number 1789
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric Acid Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'