Learn More
Hydrochloric Acid Solution 1M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, muriatic acid, spirits of salt, chlorwasserstoff, hydrochloric acid, acide chlorhydrique, hydrogen chloride, 6AMP, 57 deg.C, CHEBI:17883, Colorless, 36.45g/mol
Brand: Fisher Chemical J/4320C/90
148.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
ClH, CAS Number-7647-01-0, chloorwaterstof, anhydrous hydrochloric acid, hydrogen chloride hcl, chlorohydric acid, muriatic acid, spirits of salt, chlorwasserstoff, hydrochloric acid, acide chlorhydrique, hydrogen chloride, 6AMP, 57 deg.C, CHEBI:17883, Colorless, 36.45g/mol

Specifications
1 | |
7647-01-0 | |
57°C | |
125mbar at 20°C | |
ClH | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 |
Colorless | |
-35°C | |
Ampoule | |
6 Ampules | |
1.9 mPaS at 15°C | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.45g/mol |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Hydrochloric acid, 1M, (1N) concentrated solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2528095 | Certificate of Analysis | 25/02/2025 | J/4320C/05,J/4320/99,J/4320C/90 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.