missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 1 M HCl (1N), Honeywell Fluka™
320.75 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 L
5 L
10 L
20 L
30 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
Plastic Pail
VOLPAC™ CONTAINER
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Packaging | Plastic Bottle |
Vapor Pressure | 190 mmHg (25°C) |
Quantity | 4 x 5 L |
UN Number | UN1789 |
IUPAC Name | hydrogen chloride |
Formula Weight | 36.46 |
Grade | Reagent Ph. Eur. |
Chemical Name or Material | Hydrochloric acid solution, volumetric, 1 M HCl (1N) |
Safety and Handling
Product Identifier
- Hydrochloric acid solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'