missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 1 M HCl (1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 1 M HCl (1N)
£24.10 - £344.38
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15654940
|
Honeywell-Fluka
35328-1L |
1 L | Plastic Bottle |
£24.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15604950
|
Honeywell-Fluka
35328-5L-VP |
5 L | VOLPAC™ CONTAINER |
£128.15
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15694940
|
Honeywell-Fluka
35328-5L |
5 L | Plastic Bottle |
£100.15
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15614950
|
Honeywell-Fluka
35328-6X1L |
6 x 1 L | Plastic Bottle |
£136.83
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15644940
|
Honeywell-Fluka
35328-10L-VP |
10 L | VOLPAC™ CONTAINER |
£224.25
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15634940
|
Honeywell-Fluka
35328-10L |
10 L | N/A | ||||||
15684940
|
Honeywell-Fluka
35328-4X5L |
4 x 5 L | Plastic Bottle |
£344.38
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15664940
|
Honeywell-Fluka
35328-20L |
20 L | N/A | ||||||
15674940
|
Honeywell-Fluka
35328-30L |
30 L | Plastic Pail |
£342.51
30 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
7647-01-0 | |
190 mmHg (25°C) | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid solution |
1M | |
ClH | |
UN1789 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrochloric acid solution
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid