missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 4M in H2O, Honeywell™

Product Code. 15697310
Click to view available options
Quantity:
1 L

4 M HCl in H2O

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 7647-01-0
Concentration 4M
Vapor Pressure 190 mmHg (25°C)
Quantity 1 L
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
UN Number UN1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid solution
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Hydrochloric acid solution
Palabra de advertencia
  • Warning
Clasificación
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'