missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M HCl (0.5N), Honeywell Fluka™
95.93 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Boiling Point | >100°C |
CAS Min % | ≥90.0000%, ≥1.0000% |
Concentration | 0.5 M HCl (0.5N) |
CAS Max % | ≤100.0000%, <5.0000% |
Density | 1.010 g/cm3 |
Packaging | Poly Bottle |
Quantity | 6 x 1 L |
Linear Formula | HCl |
UN Number | UN1789 |
IUPAC Name | hydrogen chloride |
Show More |
Safety and Handling
Product Identifier
- Hydrochloric acid solution
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : hydrochloric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'