missing translation for 'onlineSavingsMsg'
Learn More

Hydrocortisone, Tocris Bioscience™

Product Code. 18084792
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Adrenal glucocorticoid; immunosuppressant

Hydrocortisone is an adrenal glucocorticoid released in the stress response. Increases glucose production by gluconeogenesis. Immunosuppressant and antidiuretic hormone. Used in protocols to generate retinal pigment epithelial (RPE) cells from PSCs.

Specifications

CAS 50-23-7
Molecular Formula C21H30O5
Quantity 50 mg
Synonym hydrocortisone, cortisol, acticort, cetacort, cortef, 17-hydroxycorticosterone, hydrocortisyl, hydrasson, cobadex, cortril
InChI Key JYGXADMDTFJGBT-VWUMJDOOSA-N
SMILES CC12CCC(=O)C=C1CCC3C2C(CC4(C3CCC4(C(=O)CO)O)C)O
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 362.466
PubChem CID 5754
ChEBI CHEBI:17650
Formula Weight 362.46
Percent Purity >98%
Chemical Name or Material Hydrocortisone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'