Learn More
Macherey-Nagel™ Hydrodex β-6TBDM GC Capillary Column
Brand:  Macherey-Nagel™ 723383.10
 View more versions of this product
Product Code. 12492760
  • £738.00 / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
0.4 mm
0.1 mm
Enantiomer Separation
10 m
1 Each
GC Capillary Column
Hydrodex β-6TBDM
230°C to 250°C
Hydrodex β-6TBDM