missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid 47-51%, Trace Metal™, for Trace Metal Analysis, Fisher Chemical
132.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
500mL
Specifications
pH | 1 |
Color | Colorless |
CAS | 7664-39-3 |
Melting Point | -35°C |
Boiling Point | 105°C |
Quantity | 500 mL |
Molecular Formula | FH |
Physical Form | Liquid |
MDL Number | MFCD00011346 |
InChI Key | KRHYYFGTRYWZRS-UHFFFAOYSA-N |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Hydrofluoric acid, solution in water
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrogen fluoride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'