missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, 48%, Certified AR for Analysis, Fisher Chemical™
£180.00 - £240.00
Chemical Identifiers
CAS | 7664-39-3 |
---|---|
Molecular Formula | FH |
Molecular Weight (g/mol) | 20.01 |
MDL Number | MFCD00011346 |
InChI Key | KRHYYFGTRYWZRS-UHFFFAOYSA-N |
Synonym | hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 |
PubChem CID | 14917 |
ChEBI | CHEBI:29228 |
SMILES | F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10418600
|
Fisher Chemical
H/1420/15 |
1 L |
£180.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402664
|
Fisher Chemical
H/1420/17 |
2.5 L |
£240.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrofluoric Acid, 48%, Certified AR for Analysis
Chemical Identifiers
7664-39-3 | |
20.01 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
14917 | |
F |
FH | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
CHEBI:29228 |
Specifications
1 | |
7664-39-3 | |
105°C | |
FH | |
MFCD00011346 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
hydrogen fluoride | |
14917 | |
20g/mol |
Colorless | |
-35°C | |
HDPE plastic bottle | |
Liquid | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
F | |
20.01 | |
CHEBI:29228 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrofluoric acid, 48% solution in water
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 1
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen fluoride