missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals

Product Code. 10370234 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10370234

Brand: Thermo Scientific Acros 423800025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7664-39-3, 7732-18-5
Melting Point -35.0°C
Boiling Point 105.0°C
Density 1.1500g/mL
Heavy Metals (as Pb) 0.5ppm max.
Packaging Plastic bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.15
Quantity 2.5 kg
Molecular Formula FH
Linear Formula HF
MDL Number MFCD00011346
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
Solubility Information Solubility in water: soluble.
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES F
IUPAC Name hydrogen fluoride
Molecular Weight (g/mol) 20.01
PubChem CID 14917
ChEBI CHEBI:29228
Formula Weight 20
Grade ACS Reagent
Chemical Name or Material Hydrofluoric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrofluoric acid, 48 to 51% solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H310-Fatal in contact with skin.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.