missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, Baker Instra-Analyzed Reagent, USP/NF Grade, For ACS analysis, J.T. Baker™
322.05 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
Specifications
CAS | 7664-39-3 |
Heavy Metals (as Pb) | max. 100ppb |
Packaging | Poly Bottle |
Residue after Ignition | max. 5ppm |
Specific Gravity | 1L = 0.9909kg |
Assay Percent Range | 48.0 to 51.0% |
Quantity | 500 mL |
Molecular Formula | FH |
Physical Form | Liquid |
MDL Number | MFCD00011346 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Signal Word
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'