missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric acid, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥40%, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | ≥40% |
Density | 1.15g/mL at 25°C |
Residue after Ignition | ≤0.0005% (as SO4) |
Vapor Pressure | 25mmHg (20°C) |
Quantity | 1 L |
UN Number | 1790 |
SMILES | F |
IUPAC Name | hydrogen fluoride |
Formula Weight | 20.01g/mol |
Grade | ACS/puriss. p.a. |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
Product Identifier
- Hydrofluoric acid
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H300 + H310 + H330-Fatal if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrogen fluoride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'