missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, Technical grade, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
5 L
6 x 1 L
Unit Size:
1 L
5 L
Pack of 6
Descrizione
- Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
- Variation in color and physical form is possible
Specifica
Specifica
| Concentration | 40 to 45% |
| Density | 1.15g/mL at 25°C |
| Vapor Pressure | 25mmHg (20°C) |
| Quantity | 5 L |
| UN Number | 1790 |
| SMILES | F |
| IUPAC Name | fluorane |
| Formula Weight | 20.01g/mol |
| Grade | Technical |
| Chemical Name or Material | Hydrofluoric Acid |
Sicurezza e movimentazione
Identificativo prodotto
- Hydrofluoric acid
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Hydrogen fluoride
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto