missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid, Technical grade, Honeywell Fluka™
Click to view available options
Quantity:
1 L
5 L
6 x 1 L
Description
- Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
- Variation in color and physical form is possible
Spécification
Concentration | 40 to 45% |
Density | 1.15g/mL at 25°C |
Vapor Pressure | 25mmHg (20°C) |
Quantity | 1 L |
UN Number | 1790 |
SMILES | F |
IUPAC Name | fluorane |
Formula Weight | 20.01g/mol |
Grade | Technical |
Chemical Name or Material | Hydrofluoric Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'