Learn More
Hydrogen chloride, 5 to 6N solution in 2-propanol, AcroSeal™, Thermo Scientific Chemicals
£28.20 - £43.60
Specifications
Chemical Name or Material | Hydrochloric acid |
---|---|
Color | Colorless to Yellow |
Name Note | 5 to 6N solution in 2-propanol |
CAS | 7647-01-0, 67-63-0 |
Concentration | 5N min. |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10172854
|
Thermo Scientific Acros
428601000 |
100 mL | AcroSeal™ Glass Bottle |
£28.20
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10764966
|
Thermo Scientific Acros
428608000 |
800 mL | AcroSeal™ Glass Bottle |
£43.60
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
5 to 6N solution in 2-propanol | |
5N min. | |
AcroSeal™ Glass Bottle | |
11°C | |
HCl | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.45 |
Colorless to Yellow | |
7647-01-0, 67-63-0 | |
0.9090g/mL | |
0.909 | |
ClH | |
Liquid | |
14, 4794 | |
Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L | |
Cl | |
36.46 | |
CHEBI:17883 |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Hydrogen chloride, 5 to 6N solution in 2-propanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Isopropyl alcohol, hydrogen chloride
RUO – Research Use Only