missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Chloride Solution, 1.0M in diethyl ether, Honeywell™ Fluka™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UN Number | UN2924 |
Density | 0.731g/mL at 25°C |
Formula Weight | 36.46g/mol |
Packaging | Glass Bottle |
Quantity | 100 mL |
Flash Point | −40°C (−40°F) |
Physical Form | Liquid |
Chemical Name or Material | Hydrogen Chloride Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'