missing translation for 'onlineSavingsMsg'
Learn More

Parker domnick hunter™ Hydrogen Generators

Research-grade hydrogen gas on demand

Brand:  Parker domnick hunter™ 40H

 View more versions of this product

Product Code. 12965981

  • £10900.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: One desiccant cartridge, one deionizer cartridge and drain tube.
This item is not returnable. View return policy

Description

Description

Provide a safe and economical alternative to high-pressure gas cylinders and can be easily installed on the lab bench. Ideal for carrier and fuel gas applications and suitable for use with any laboratory instrument requiring a supply of ultra-high-purity hydrogen. Eliminate interruptions in analyses to change cylinders. Three models available with 160, 250 and 500cc/min. outlet flowrates.

  • LCD display shows system status and diagnostic capabilities
  • Automatic leak detection
  • Integrated microprocessor control
  • Low water and poor quality water detection and alarms
  • Built-in switch-mode power supply unit accepts electricity supplied between 100 and 230V and 50 to 60Hz

Products in this group have a universal power supply and can be used with any voltage 100-230V 50/60Hz

  • Produce a continuous stream of research-grade hydrogen gas from deionized water using a proprietary ion exchange membrane and drying cartridge
  • Using hydrogen as a GC carrier gas increases speed and sensitivity compared to helium, decreasing analysis time without discernible loss of resolution

Order Info

  • Additional cartridges and bags sold separately
  • Connector: 0.12 in. Swagelok™

Certifications

CE, CSA and UL compliant and listed

Services

Full two-year warranty

Specifications

Specifications

CE, CSA, UL
34.2 cm
17.9 in.
5°C to 40°C
90 to 264 V
13.5 in.
40H
0 to 100 psi
2 Years
Hydrogen Fuel Gas Generator
17.2 in.
250 mL/min.
45.6 cm
0.125 in. compression fitting
23 kg
90 to 264V 50/60Hz
99.9995%
185w
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers