Learn More
hydrogen hexachloroplatinate(IV) hydrate, ACS reagent, Thermo Scientific Chemicals
£76.28 - £1229.12
Chemical Identifiers
CAS | 26023-84-7 |
---|---|
Molecular Formula | Cl6H2Pt |
Molecular Weight (g/mol) | 409.80 |
MDL Number | MFCD00149909 |
InChI Key | ZKOQTCCVSRSECD-UHFFFAOYSA-J |
Synonym | Hexachloroplatinic acid hydrate, Platinic chloride hydrate |
IUPAC Name | dihydrogen hexachloroplatinumtetrakis(ylium) |
SMILES | [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10627751
|
Thermo Scientific Acros
405010010 |
1 g | Glass bottle |
£76.28
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10106990
|
Thermo Scientific Acros
405010050 |
5 g | Glass bottle |
£258.29
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904371
|
Thermo Scientific Acros
405010250 |
25 g | Glass bottle |
£1229.12
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
26023-84-7 | |
409.80 | |
ZKOQTCCVSRSECD-UHFFFAOYSA-J | |
dihydrogen hexachloroplatinumtetrakis(ylium) |
Cl6H2Pt | |
MFCD00149909 | |
Hexachloroplatinic acid hydrate, Platinic chloride hydrate | |
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl |
Specifications
60.0°C | |
Chunks and/or Powder | |
H2PtCl6·xH2O | |
Hexachloroplatinic acid hydrate, Platinic chloride hydrate | |
ZKOQTCCVSRSECD-UHFFFAOYSA-J | |
dihydrogen hexachloroplatinumtetrakis(ylium) | |
409.82 | |
ACS Reagent |
26023-84-7 | |
Cl6H2Pt | |
MFCD00149909 | |
(in alcohol) Passes test | |
[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl | |
409.80 | |
≥37.50% (Pt) | |
Hydrogen hexachloroplatinate(IV) hydrate |
Safety and Handling
GHS H Statement
May be corrosive to metals.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.