missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, 0.3, ULTREX™ II Ultrapure Reagent, J.T. Baker™
399.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
450 mL
Packaging:
HDPE Pressure Bottom Poly Bottle
Description
- For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory,Research,or Manufacturing Use
Specifications
CAS | 7722-84-1 |
Packaging | HDPE Pressure Bottom Poly Bottle |
Molecular Formula | H2O2 |
Quantity | 450 mL |
Specific Gravity | 1L = 1.11kg |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
SMILES | OO |
IUPAC Name | hydrogen peroxide |
Molecular Weight (g/mol) | 34.014 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS03_Oxidiser.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Danger
- Acute toxicity Category 4
- Oxidising liquid Category 1
- Skin corrosion/irritation Category 1 A
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- P283-Wear fire/flame resistant/retardant clothing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'