missing translation for 'onlineSavingsMsg'
Learn More

hydrogen peroxide, 3 wt.% solution in water, stabilized

Product Code. 10081254 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
100mL
25mL
This item is not returnable. View return policy

Product Code. 10081254

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1, 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 100.0°C
Linear Formula HOOH
Density 1.0100g/mL
Merck Index 15, 4835
Solubility Information Solubility in water: soluble.
Formula Weight 34
Packaging Plastic bottle
Specific Gravity 1.01
Quantity 25 mL
Chemical Name or Material Hydrogen peroxide, 3 wt.% solution in water, stabilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.