missing translation for 'onlineSavingsMsg'
Learn More
Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Certified AR for Analysis, Fisher Chemical™ Product Code.: 10736291

Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Certified AR for Analysis, Fisher Chemical™

Product Code. 10736291
1 L, HDPE plastic bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10736291

Brand: Fisher Chemical H/1800/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

H2O2, CAS Number-7722-84-1, albone, hioxyl, hydrogen dioxide, inhibine, interox, kastone, oxydol, perhydrol, peroxaan, superoxol, 1L, 30-32%w/w, 108 deg.C, CHEBI:16240, Colorless, MHAJPDPJQMAIIY-UHFFFAOYSA-N, hydrogen peroxide, -33 deg.C, 11333, 34.014, HDPE plastic bottle, 3.3, Liquid, 784, OO, 1.1

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number 11333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 108°C
Melting Point -33°C
Assay Percent Range 30-32%w/w
Percent Purity 30 to 32%
Packaging HDPE plastic bottle
Vapor Density 1.1
pH 3.3
Quantity 1 L
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide 20-35wt% solution in water
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen peroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.