missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide 30% (w/v) (100 Volumes), Extra Pure SLR, Fisher Chemical™
£68.95 - £133.00
Chemical Identifiers
CAS | 7722-84-1 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
MDL Number | 11333 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10687022
|
Fisher Chemical
H/1750/15 |
1 L |
£68.95
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10121810
|
Fisher Chemical
H/1750/17 |
2.5 L |
£133.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7722-84-1 | |
34.014 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
H2O2 | |
11333 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
Specifications
Colorless | |
108°C | |
H2O2 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
1.1 | |
Liquid |
7722-84-1 | |
-33°C | |
11333 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
HDPE plastic bottle | |
3.3 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Hydrogen peroxide 20-35wt% solution in water
Signal Word
- Danger
Hazard Category
- Oxidising liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen peroxide