missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide (30% in Water), Fisher BioReagents
Click to view available options
Quantity:
500 mL
Description
Hydrogen Peroxide is used as an antiseptic and for decomposition of inorganic and organic samples.

Specifications
Specifications
Color | Colorless |
CAS | 7722-84-1, 7732-18-5 |
Boiling Point | 108°C |
Melting Point | -33°C |
Assay Percent Range | (as H2O2, %wt): 29.0 to 32.0% |
Molecular Formula | H2O2 |
Density | 1.11g/cm³ |
Merck Index | 15, 4835 |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Show More |
Safety and Handling



Product Identifier
- Hydrogen peroxide 20-35wt% solution in water
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Oxidising liquid Category 2
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrogen peroxide
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrogen Peroxide (30% in Water), Fisher BioReagents >
Spot an opportunity for improvement?Share a Content Correction