missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution 30-32%, Optima™, for Ultra Trace Analysis, Fisher Chemical
627.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
Specifications
Color | Undesignated |
CAS | 7722-84-1 |
Boiling Point | 108°C |
Melting Point | -33°C |
Molecular Formula | H2O2 |
MDL Number | 11333 |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
SMILES | OO |
IUPAC Name | hydrogen peroxide |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS03_Oxidiser.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Hydrogen peroxide 30-32%, Optima for Ultra traces analysis
- Danger
- Oxidising liquid Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Hydrogen peroxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'