missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T. Baker™
£87.00 - £464.00
Chemical Identifiers
CAS | 7722-84-1 |
---|---|
Molecular Formula | H2O2 |
Molecular Weight (g/mol) | 34.014 |
InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
Synonym | oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone |
PubChem CID | 784 |
ChEBI | CHEBI:16240 |
IUPAC Name | hydrogen peroxide |
SMILES | OO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15567984
|
Avantor J.T.Baker
2182-01 |
500 mL | Narrow Mouth Amber Glass Bottle | N/A | ||||||
15577984
|
Avantor J.T.Baker
2182-03 |
4 L | Narrow Mouth Amber Glass Bottle |
£464.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Chemical Identifiers
7722-84-1 | |
34.014 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
CHEBI:16240 | |
OO |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
784 | |
hydrogen peroxide |
Specifications
7722-84-1 | |
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone | |
OO | |
34.014 | |
CHEBI:16240 | |
2.5 to 3.5% | |
1L = 1.00kg |
H2O2 | |
MHAJPDPJQMAIIY-UHFFFAOYSA-N | |
hydrogen peroxide | |
784 | |
34.01 | |
USP | |
Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hydrogen Peroxide, Topical Solution, Contains 0.05% max. Acetophenetidin as a Preservative, 2.5 to 3.5%, U.S.P., J.T. Baker™