missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Phthalate - C6H4(COO)2H for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£50.50 - £104.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12972125
|
Fisher Chemical
A 020 100 |
100 mL | Glass Bottle |
£50.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12982125
|
Fisher Chemical
A 020 250 |
250 mL | Glass Bottle |
£70.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12992125
|
Fisher Chemical
A 020 500 |
500 mL | Glass Bottle |
£104.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
C6H4(COO)2H- |
Specifications
1g/L | |
C6H4(COO)2H- | |
Liquid |
Single-component Standard Solution | |
H2O | |
Hydrogen Phthalate |
Safety and Handling
missing translation for 'shelfLife' : 24 Months. (12 Months after opening)