missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Potassium ATPase Beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-32035
This item is not returnable.
View return policy
Description
Hydrogen Potassium ATPase Beta Polyclonal specifically detects Hydrogen Potassium ATPase Beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Hydrogen Potassium ATPase Beta | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
P51164 | |
ATP4B | |
This antibody was developed against a recombinant protein corresponding to amino acids: TPDYQDQLRSPGVTLRPDVYGEKGLEIVYNVSDNRTWADLTQTLHAFLAGYSPAAQEDSINCTSEQYFFQESFRAPNHTKFSCKFTADML | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain | |
Rabbit | |
Affinity Purified | |
RUO | |
496 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction