missing translation for 'onlineSavingsMsg'
Learn More

BUSE Scientific Hydrogen Sulfide Gas

Brand:  BUSE Scientific LBHYDS25

Code : 22

Additional Details : CAS Number : 7783-06-4 Weight : 0.22700kg Transport : UN number : 1053 Chem class : 2.3

Product Code. 12904940

  • £376.88 / 227g
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

Specifications

Specifications

227 g
Hydrogen Sulfide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers