missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Packaging | Glass bottle |
Molecular Formula | AuCl4H |
Quantity | 1 g |
UN Number | UN3260 |
Sensitivity | Hygroscopic; Light sensitive |
Synonym | Tetrachloroauric(III) acid,Chloroauric acid |
Solubility Information | Soluble in water. Insoluble in ether. |
InChI Key | RFWCEGCWRSNAGT-UHFFFAOYSA-K |
Molecular Weight (g/mol) | 339.77 |
Formula Weight | 339.79 (anhydrous) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'