Learn More
Brannan™ Hydrometer No. 1 Lactometer For Dairy Testing Glass Without Works Certificate
Hydrometer No. 1 Lactometer For Dairy Testing Glass Without Works Certificate, 1.025 to 1.035 x 0.0002 150mm length Brannan
Brand: Brannan™ 10/602/0
This item is not returnable.
View return policy
Description
Hydrometer No. 1 Lactometer For Dairy Testing Glass Without Works Certificate, 1.025 to 1.035 x 0.0002 150mm length Brannan
1.025 to 1.035 x 0.0002 150mm length BrannanSpecifications
Hydrometer | |
150 mm | |
Specific Gravity Measurement |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Brannan™ Hydrometer No. 1 Lactometer For Dairy Testing Glass Without Works Certificate >
Spot an opportunity for improvement?Share a Content Correction