missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Hydrophilic Dialysis Tubing
Ultra high purity separation dialysis membranes
£475.00
Specifications
Product Type | Dialysis Tubing |
---|---|
Diameter (Metric) | 10 mm |
Length (Metric) | 10 m |
Capacity (Metric) | 0.79 mL/cm |
Quantity | 1 Each |
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
11327030
|
Spectrum™ Labs
131450 |
300 kDa |
£475.00
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12787516
|
Spectrum™ Labs
131486 |
1 kDa |
£475.00
Each |
Estimated Shipment: 18-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
An ultra high purity separation dialysis membrane. The Spectra/Por™ Cellulose Ester (CE) dialysis membranes offer an excellent Molecular Weight Cut Off (MWCO) selection for separation of ionic species as well as macromolecules. The advance manufacturing process provides CE dialysis membranes with an extremely high purity and features a biological inert surface to yield an optimum sample recovery. Symmetric tubular CE membranes are supplied in form of non-sterile, wet tubular membrane.Chemical resistance
Variables in temperature, concentrations, durations of exposure and other factors may affect the use of the product. CE membranes are sensitive to organic solvents at concentrations exceeding 5% in water. Strong polar solvents such as acetone, Methyl Ethyl Ketone (MEK) or Dioxane will irreparably damage the membranes. Lower alcohols such as Methanol, and Isopropanol can be used with CE membranes at short exposure or low concentrations.
- Hydrophilic and uniform molecular porous dialysis membranes
- Nine MWCOs: 100D to 500D, 500D to 1,000D, 3, 5kD to 5kD, 8kD to 10kD, 20kD, 50kD, 100kD, 300kD and 1,000kD
- Four flat widths: 10mm, 16mm, 24mm, 33mm
- No special chemical pre-cleaning required
- Non-sterile Biotech tubular CE membranes
- Supplied wet in 0.1% Sodium Azide
- Supplied in 10m length (wet)
Specifications
Dialysis Tubing | |
10 m | |
1 Each |
10 mm | |
0.79 mL/cm |