missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Description
Hydroxyapatite nanoparticles is used for polishing and dental powders, porcelain and pottery, and enameling. It is also utilized to engineer new biomaterials for applications such as bone grafts and fillers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Content And Storage | 2°C to 8°C |
Format | Liquid dispersion |
Quantity | 25 mL |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'